Få mere at vide
Pentanedioic-3,3-d2 Acid, CDN
Artikelnummer. 30320037
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30320037 0.25 g -
30309186 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320037 Leverandør CDN Leverandørnr. CDND59430.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pentanedioic-3,3-D2 Acid
CAS 43087-19-0
Molekylær formel C5 D2 H6 O4
Mængde 0.25 g
Synonym Pentanedioic-3,3-d2 acid (9 CI, ACI), Glutaric-3,3-d2 acid, Pentanedioic acid-3,3-d2, Glutaric acid-3,3-d2, Glutaric-d2 acid, Glutaric acid-d2, Pentanedioic-3,3-D2 acid, Pentanedioic-d2 acid
InChI formel InChI=1 S/C5H8O4/c6-4(7)2-1-3-5(8)9/h1-3H2,(H,6,7)(H,8,9)/i1D2
SMIL [2 H]C([2 H])(CC(=O)O)CC(=O)O
IUPAC navn 3,3-dideuteriopentanedioic acid
Molekylvægt (g/mol) 134.127
Formel vægt 134.055 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity