Learn More
Pentane-1-sulfonic acid sodium salt, LiChropur™ EMD-Millipore
Product Code. 15825890
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass
Unit Size:
25g
Product Code. Quantity Packaging Unit Size
15825890 25 g Glass 25g
1 options
Product Code. 15825890 Supplier Supelco Supplier No. 1183040025
 Restricted Item
Forventet forsendelse: 18-11-2026
Ion pair reagents for analytical HPLC LiChropur
TRUSTED_SUSTAINABILITY
Specifications
CAS 22767-49-3
Density 390kg/m3
Packaging Glass
Molecular Formula C5H11NaO3S
MDL Number MFCD00007541,MFCD00149548
Quantity 25 g
Synonym sodium 1-pentanesulfonate, sodium pentane-1-sulfonate, sodium pentanesulfonate, 1-pentanesulfonic acid sodium salt, 1-pentanesulfonic acid, sodium salt, 1-pentylsodiumsulfonate, ipc-alks-5, 1-pentanesulfonate, sodium, pentanesulfonic acid na-salt, pentanesulfonic acidsodiumsalt
InChI Key ROBLTDOHDSGGDT-UHFFFAOYSA-M
SMILES [Na+].CCCCCS([O-])(=O)=O
IUPAC Name sodium pentane-1-sulfonate
Molecular Weight (g/mol) 174.19
PubChem CID 23664617
Formula Weight 174.2
Percent Purity ≥99%
Grade Not Available
Physical Form Solid
Chemical Name or Material Pentane-1-sulfonic acid sodium salt