Få mere at vide
Pentamidine Diisethionate, TRC
Artikelnummer. 17984875
Skift visning
Mængde:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17984875 5 g
17954255 1 g
17964405 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17984875 Leverandør Toronto Research Chemicals Leverandørnr. TRCP2725015G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 140-64-7
Molekylær formel C19 H24 N4 O2 . 2 C2 H6 O4 S
Molekylvægt (g/mol) 592.68
Synonym Pentamidine diisetionate, 4,4'-[Pentane-1,5-diylbis(oxy)]dibenzamidine di(2-hydroxyethanesulfonate), Ethanesulfonic acid, 2-hydroxy-, compd. with 4,4'-[1,5-pentanediylbis(oxy)]bis[benzenecarboximidamide] (2:1), Benzamidine, 4,4'-(pentamethylenedioxy)di-, diisethionate (6CI), Ethanesulfonic acid, 2-hydroxy-, compd. with 4,4'-(pentamethylenedioxy)dibenzamidine (2:1) (8CI), Benzenecarboximidamide, 4,4'-[1,5-pentanediylbis(oxy)]bis-, bis(2-hydroxyethanesulfonate) (9CI), 1,5-Bis(4-amidinophenoxy)pentane diisethionate, 2512RP, 4,4'-Diamidino-α,ω-diphenoxypentane diisethionate, 4,4'-Diamidinodiphenoxypentane di(β-hydroxyethanesulfonate), Aeropent, Banambax, Diamidine, Lomidin, Lomidine, M & B 800, NebuPent, Pentacarinat, Pentam, Pentam 300, Pentamidine diisethionate, Pentamidine isethionate, Pentamidine isetionate, Pneumopent, RP 2512, WR 4931, p,p'-(Pentamethylenedioxy)dibenzamidine bis(β-hydroxyethanesulfonate)
IUPAC navn 4-[5-(4-carbamimidoylphenoxy)pentoxy]benzenecarboximidamide;2-hydroxyethanesulfonic acid
SMIL NC(=N)c1ccc(OCCCCCOc2ccc(cc2)C(=N)N)cc1.OCCS(=O)(=O)O.OCCS(=O)(=O)O
Tekniske data
Kemisk navn eller materiale Pentamidine Diisetionate
Mængde 5 g
InChI formel InChI=1S/C19H24N4O2.2C2H6O4S/c20-18(21)14-4-8-16(9-5-14)24-12-2-1-3-13-25-17-10-6-15(7-11-17)19(22)23;2*3-1-2-7(4,5)6/h4-11H,1-3,12-13H2,(H3,20,21)(H3,22,23);2*3H,1-2H2,(H,4,5,6)
Formel vægt 592.1873