missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl pyrimidine-5-carboxylate, 97%, Thermo Scientific™

Catalog Number p-3541160
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
921938-48-9
C11H3F5N2O2
290.15
MFCD09702344
TZPLYNSLIVPKAH-UHFFFAOYSA-N
pentafluorophenyl pyrimidine-5-carboxylate, perfluorophenyl pyrimidine-5-carboxylate, 2,3,4,5,6-pentafluorophenyl pyrimidine-5-carboxylate, 2,3,4,5,6-pentakis fluoranyl phenyl pyrimidine-5-carboxylate, 5-pyrimidinecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester
24229455
(2,3,4,5,6-pentafluorophenyl) pyrimidine-5-carboxylate
FC1=C(F)C(F)=C(OC(=O)C2=CN=CN=C2)C(F)=C1F
This item is not returnable. View return policy
921938-48-9
C11H3F5N2O2
290.15
MFCD09702344
TZPLYNSLIVPKAH-UHFFFAOYSA-N
pentafluorophenyl pyrimidine-5-carboxylate, perfluorophenyl pyrimidine-5-carboxylate, 2,3,4,5,6-pentafluorophenyl pyrimidine-5-carboxylate, 2,3,4,5,6-pentakis fluoranyl phenyl pyrimidine-5-carboxylate, 5-pyrimidinecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester
24229455
(2,3,4,5,6-pentafluorophenyl) pyrimidine-5-carboxylate
FC1=C(F)C(F)=C(OC(=O)C2=CN=CN=C2)C(F)=C1F
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 290.15
Percent Purity 97%
Chemical Name or Material Pentafluorophenyl pyrimidine-5-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'