Learn More
Pentaethylenehexamine, Sigma-Aldrich™
Specifications
Specifications
| CAS | 4067-16-7 |
| Melting Point | -35°C |
| Density | 1.003 g/cm3 at 20°C |
| MDL Number | MFCD00008167 |
| Quantity | 25 mL |
| Synonym | Pentaethylenehexamine, 3,6,9,12-Tetraazatetradecane-1,14-diamine, Pentaethylenehexamine |
| Molecular Weight (g/mol) | 232.37 |
| Grade | Research |