Få mere at vide
Pentaerythritol-d8, TRC
Artikelnummer. 30427292
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30427292 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30427292 Leverandør Toronto Research Chemicals Leverandørnr. TRCP2699521MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pentaerythritol-d8
CAS 82414-60-6
Molekylær formel C5 2H8 H4 O4
Mængde 1 mg
Synonym 1,3-Propane-1,1,3,3-d4-diol','2,2-bis(hydroxymethyl-d2)- (9CI), Pentaerythritol-d8, 2,2-Bis(hydroxymethyl-d2)-1,3-propanediol-1,1,3,3-d4, 2,2-Bis(hydroxymethyl)-1,3-propanediol-d8, Tetra(hydroxymethyl-d2)methane, Tetrakis(hydroxymethyl-d2)methane, Pentaerythritol-D8
InChI formel InChI=1S/C5H12O4/c6-1-5(2-7,3-8)4-9/h6-9H,1-4H2/i1D2,2D2,3D2,6D,7D
SMIL [2H]OC([2H])([2H])C(CO)(C([2H])([2H])O)C([2H])([2H])O[2H]
IUPAC navn 1,1-dideuterio-2,2-bis[dideuterio(deuteriooxy)methyl]propane-1,3-diol
Molekylvægt (g/mol) 144.2
Formel vægt 144.1238