Få mere at vide
Pentadecanal, TRC
Artikelnummer. 30419235
Skift visning
Mængde:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30419235 5 g
30416434 250 mg
30409850 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30419235 Leverandør Toronto Research Chemicals Leverandørnr. TRCP2271005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2765-11-9
Molekylær formel C15H30O
Molekylvægt (g/mol) 226.4
Synonym 1-Pentadecanal, Pentadecaldehyde, n-Pentadecanal
IUPAC navn pentadecanal
SMIL CCCCCCCCCCCCCCC=O
Tekniske data
Kemisk navn eller materiale Pentadecanal
Mængde 5 g
InChI formel InChI=1S/C15H30O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16/h15H,2-14H2,1H3
Formel vægt 226.2297