Få mere at vide
Pentacynium Bis(methyl Sulfate), LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Pentacynium Bis(methyl Sulfate) |
| CAS | 3810-83-1 |
| Analyt- eller komponentnavne | Pentacynium Bis(methyl Sulfate) |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C27 H39 N3 O . 2 C H3 O4 S |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Benzenepentanaminium, ε-cyano-N,N-dimethyl-N-[2-(4-methylmorpholinium-4-yl)ethyl]-ε-phenyl-, methyl sulfate (1:2), 4-[2-[(5-Cyano-5,5-diphenylpentyl)methylamino]ethyl]-4-methylmorpholinium methyl sulfate methosulfate (7CI), Morpholinium, 4-[2-[(5-cyano-5,5-diphenylpentyl)dimethylammonio]ethyl]-4-methyl-, bis(methyl sulfate) (8CI,9CI), N1-(5-Cyano-5,5-diphenylpentyl)-N1,N1,N2-trimethylethylene-1-ammonium-2-morpholinium bismethylsulfate, Pentacynium bis(methyl sulfate), Pentacyone mesylate, Presidal bis(methyl sulfate) |
| InChI formel | InChI=1S/C27H39N3O.2CH4O4S/c1-29(2,18-19-30(3)20-22-31-23-21-30)17-11-10-16-27(24-28,25-12-6-4-7-13-25)26-14-8-5-9-15-26;2*1-5-6(2,3)4/h4-9,12-15H,10-11,16-23H2,1-3H3;2*1H3,(H,2,3,4)/q+2;;/p-2 |
| Løsningstype | Neat |
| SMIL | COS(=O)(=O)[O-].COS(=O)(=O)[O-].C[N+](C)(CCCCC(C#N)(c1ccccc1)c2ccccc2)CC[N+]3(C)CCOCC3 |
| Vis mere |