Learn More
Pentachlorothioanisole, TRC
Product Code. 30011026
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011026 10 g 10g
17923835 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30011026 Supplier Toronto Research Chemicals Supplier No. TRCP23817010G
 Restricted Item
Forventet forsendelse: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1825-19-0
Molecular Formula C7 H3 Cl5 S
Molecular Weight (g/mol) 296.43
Synonym Benzene, pentachloro(methylthio)- (9CI), Sulfide, methyl pentachlorophenyl (6CI,7CI,8CI), 1,2,3,4,5-Pentachloro-6-(methylthio)benzene, Methyl pentachlorophenyl sulfide, Methylthiopentachlorobenzene, PCTA, Pentachloro(methylthio)benzene, Pentachlorophenyl methyl sulfide, Pentachlorothioanisole
IUPAC Name 1,2,3,4,5-pentachloro-6-methylsulfanylbenzene
SMILES CSc1c(Cl)c(Cl)c(Cl)c(Cl)c1Cl
Specifications
Quantity 10 g
InChI Formula InChI=1S/C7H3Cl5S/c1-13-7-5(11)3(9)2(8)4(10)6(7)12/h1H3
Formula Weight 293.8398
Chemical Name or Material Methyl-pentachlorophenyl sulfide