Få mere at vide
Pentacene-d14 (Major), TRC
Artikelnummer. 30417363
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30417363 1 mg
30420197 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30417363 Leverandør Toronto Research Chemicals Leverandørnr. TRCP2377721MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 63912-16-3
Molekylær formel C22D14
Molekylvægt (g/mol) 292.43
Synonym 2,3:6,7-Dibenzanthracene-d14, Benzo[b]naphthacene-d14, Dibenz[b,i]anthracene-d14, NSC 90784-d14, lin-Dibenzanthracene-d14, lin-Naphthoanthracene-d14
SMIL [2H]C1=C([2H])C([2H])=C([2H])C2=C1C([2H])=C(C([2H])=C(C([2H])=C(C([2H])=C([2H])C([2H])=C3[2H])C3=C4[2H])C4=C5[2H])C5=C2[2H]
Tekniske data
Kemisk navn eller materiale Pentacene-d14
Mængde 1 mg
InChI formel InChI=1S/C22H14/c1-2-6-16-10-20-14-22-12-18-8-4-3-7-17(18)11-21(22)13-19(20)9-15(16)5-1/h1-14H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D,13D,14D
Formel vægt 292.1974