missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Pentacarbonyliron, 99.5%

As a catalyst in organic reactions, and as carbonyl iron for high-frequency coils

Brand:  Alfa Aesar™ 031113.A1

Additional Details : CAS Number : 13463-40-6 Weight : 1.00000kg Transport : UN number : 1994 Chem class : 6.1 Pack group : I

Quantity 1kg

 View more versions of this product

Product Code. 15471177

  • 2072.85 DKK / 1kg


Estimated Shipment date
from Supplier 20-02-2019
Add to basket

Specifications

Chemical Identifiers

13463-40-6
MFCD00011001
Iron pentacarbonyl; Iron carbonyl

Specifications

Pentacarbonyliron
−15° (5°F)
1.5196
103°C (200°C (decomposition))
-20°C
13463-40-6
MFCD00011001
14,5099
Soluble in nickel tetracarbonyl and organic solvents; Insoluble in water
Liquid
1.457g/mL
Pungent
Air & Light Sensitive
Red
1kg
Fe(CO)5
UN1994
Iron pentacarbonyl; Iron carbonyl
195.9
99.5%
Videos
SDS