missing translation for 'onlineSavingsMsg'
Få mere at vide

Pentabromoethylbenzene, TRC

Product Code. 30421976
Change view
Click to view available options
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30421976 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30421976 Supplier Toronto Research Chemicals Supplier No. TRCP220270250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2,3,4,5,6-Pentabromoethylbenzene
CAS 85-22-3
Renhedsgrad noter HPLC
Molekylær formel C8 H5 Br5
Mængde 250 mg
Synonym 2,3,4,5,6-Pentabromoethylbenzene, Benzene','1,2,3,4,5-pentabromo-6-ethyl-, Benzene','pentabromoethyl- (6CI,7CI,8CI,9CI), 1,2,3,4,5-Pentabromo-6-ethylbenzene, 2,3,4,5,6-Pentabromoethylbenzene, EB 80, PBEB, Pentabromoethylbenzene
InChI formel InChI=1S/C8H5Br5/c1-2-3-4(9)6(11)8(13)7(12)5(3)10/h2H2,1H3
SMIL CCc1c(Br)c(Br)c(Br)c(Br)c1Br
IUPAC navn 1,2,3,4,5-pentabromo-6-ethylbenzene
Molekylvægt (g/mol) 500.65
Formel vægt 495.6308
Procent renhed >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.