missing translation for 'onlineSavingsMsg'
Få mere at vide

Pentabromobenzyl Acrylate, TRC

Artikelnummer. 30415777
Change view
Click to view available options
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30415777 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30415777 Supplier Toronto Research Chemicals Supplier No. TRCP227120500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Pentabromobenzyl acrylate
CAS 59447-55-1
Molekylær formel C10 H5 Br5 O2
Mængde 500 mg
Synonym 2-Propenoic acid','(2,3,4,5,6-pentabromophenyl)methyl ester, 2-Propenoic acid','(pentabromophenyl)methyl ester (9CI), 2,3,4,5,6-Pentabromobenzyl acrylate, Actimer FR 1025M, FR 1025M, PBBA
InChI formel InChI=1S/C10H5Br5O2/c1-2-5(16)17-3-4-6(11)8(13)10(15)9(14)7(4)12/h2H,1,3H2
SMIL Brc1c(Br)c(Br)c(COC(=O)C=C)c(Br)c1Br
IUPAC navn (2,3,4,5,6-pentabromophenyl)methyl prop-2-enoate
Molekylvægt (g/mol) 556.6655
Formel vægt 551.6206
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.