missing translation for 'onlineSavingsMsg'
Få mere at vide

Penicillin V Potassium Salt, TRC

Artikelnummer. 30132718
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132718 25 g
17922895 100 mg
17983325 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30132718 Leverandør Toronto Research Chemicals Leverandørnr. TRCP22350025G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 132-98-9
Molekylær formel C16 H17 N2 O5 S . K
Molekylvægt (g/mol) 388.48
Synonym Phenoxymethylpenicillin Potassium, (2S,5R,6R)-3,3-Dimethyl-7-oxo-6-[(2-phenoxyacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid potassium salt (1:1), 3,3-Dimethyl-7-oxo-6-(2-phenoxyacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid monopotassium salt, (2S,5R,6R)-3,3-Dimethyl-7-oxo-6-(2-phenoxyacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid monopotassium salt, [2S-(2α,5α,6β)]-3,3-Dimethyl-7-oxo-6-(2-phenoxyacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid monopotassium salt, Abbocillin VK, Acipen, Anapenil, Antibiocin, Apo-Pen VK, Apsin VK, Arcacil, Arcasin, Asillin, Beapen, Beepen Vk, Beromycin, Beromycin (penicillin), Beromycin 400, Betapen V, Betapen Vk, Calciopen K, Cilacil, Cilicaine VK, Cliacil, Compocillin VK, DQV-K, Distakaps V-K, Distaquaine V-K, Dowpen V-K, DuraPenicillin, Fenocin, Fenocin Forte, Fenospen, Fenoxcillin, Fenoxypen, Icipen, Isocillin, Ispenoral, Kavepenin, L.P.V., Ledercillin Vk, Len VK, Megacilina Oral, Megacillin-oral, Microcillin VK, Micropen VK, Milcopen, Monocillin E, Nadopen V, Newcillin, Novo VK, Novopen VK, Ocillin VK, Oracil VK, Oracillin VK, Orapen, Orvek, Ospen 250, Ospen KV, Ospeneff, P.V.O., PVK, Pedipen, Pen V-K, Pen-V, Pen-Vee K Powder, Pen-Vee-K, Pen-Vi-K, Penadur VK Mega, Penagen, Penapar VK, Pencompren, Penicillin V potassium, Penicillin V potassium salt, Penicillin VK, Penivoral, Penoxilin, Pentabs, Pentacillin, Pentranex, Penvikal, Penvisil, Pfizerpen Vk, Phenoxymethylpenicillin potassium, Phenoxymethylpenicillin potassium salt, Potassium 2,2-dimethyl-6α-phenoxyacetamidopenam-3α-carboxylate, Potassium penicillin V, Potassium penicillin V salt, Potassium phenoxymethylpenicillin, Potassium phenoxymethylpenicillinate, Primcillin, Qidpen VK, Rafapen V-K, Robicillin VK, Rocilin, Rocillin VK, Roscopenin, SK-Penicillin VK, Servipen V, Stabilin V-K, Stabillin VK syrup 125, Stabillin VK syrup 62.5, Sumapen VK, Suspen, Trepopen, Trepopen VK, Uticillin Vk, V-Cil K, V-Cillin K, V-Kal K, V-Pen, V-Penicillin Kalium, Vamosyn, Veetids, Vepen, Vepicombin
IUPAC navn potassium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenoxyacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMIL [K+].CC1(C)S[C@@H]2[C@H](NC(=O)COc3ccccc3)C(=O)N2[C@H]1C(=O)[O-]

Tekniske data

Kemisk navn eller materiale Phenoxymethylpenicillin Potassium
Renhedsgrad noter HPLC
Mængde 25 g
InChI formel InChI=1S/C16H18N2O5S.K/c1-16(2)12(15(21)22)18-13(20)11(14(18)24-16)17-10(19)8-23-9-6-4-3-5-7-9;/h3-7,11-12,14H,8H2,1-2H3,(H,17,19)(H,21,22);/q;+1/p-1/t11-,12+,14-;/m1./s1
Formel vægt 388.0495
Procent renhed >95%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.