missing translation for 'onlineSavingsMsg'
Learn More

Penicillin V potassium salt, 100 mg/mL in distilled water, sterile-filtered, Thermo Scientific™

Catalog Number p-7122696
Click to view available options
Quantity:
50 mL
100 mL
This item is not returnable. View return policy
132-98-9
C16H17KN2O5S
388.48
MFCD00051771
HCTVWSOKIJULET-LQDWTQKMSA-M
131673927
(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenoxyacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;potassium
[K+].CC1(C)S[C@@H]2[C@H](NC(=O)COC3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O
This item is not returnable. View return policy
132-98-9
C16H17KN2O5S
388.48
MFCD00051771
HCTVWSOKIJULET-LQDWTQKMSA-M
131673927
(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenoxyacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;potassium
[K+].CC1(C)S[C@@H]2[C@H](NC(=O)COC3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Type Penicillin V Potassium Salt
Physical Form Liquid
Concentration 100mg/mL in distilled water
Quantity 50 mL
Chemical Name or Material Penicillin V potassium salt, sterile-filtered
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'