Få mere at vide
Penicillin V Beta-Sulfoxide-d3, TRC
Artikelnummer. 30425643
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30425643 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30425643 Leverandør Toronto Research Chemicals Leverandørnr. TRCP2235071MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Penicillin V beta-Sulfoxide-d3
CAS 148680-07-3
Molekylær formel C16 D3 H15 N2 O6 S
Mængde 1 mg
Synonym 6-Phenoxyacetamidopenicillinic Acid-d3 1beta-Oxide, Penicillin-d3 V S-Oxide, [2S-(2alpha,4beta,5alpha,6beta)]-3,3-Dimethyl-7-oxo-6-[(phenoxyacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid-d3 4-Oxide, (2S,4S,5R,6R)-3,3-Dimethyl-7-oxo-6-[(2-phenoxyacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid-d3 4-Oxide
InChI formel InChI=1S/C16H18N2O6S/c1-16(2)12(15(21)22)18-13(20)11(14(18)25(16)23)17-10(19)8-24-9-6-4-3-5-7-9/h3-7,11-12,14H,8H2,1-2H3,(H,17,19)(H,21,22)/t11-,12+,14-,25-/m1/s1/i1D3/t11-,12+,14-,16-,25-
SMIL [2H]C([2H])([2H])[C@@]1(C)[C@@H](N2[C@@H]([C@H](NC(=O)COc3ccccc3)C2=O)[S@+]1[O-])C(=O)O
IUPAC navn (2S,3S,4S,5R,6R)-3-methyl-4-oxido-7-oxo-6-[(2-phenoxyacetyl)amino]-3-(trideuteriomethyl)-4-thionia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Molekylvægt (g/mol) 369.407
Formel vægt 369.107