Få mere at vide
Penicillin G-D7 N-Ethylpiperidinium Salt, TRC
Artikelnummer. 30421316
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30421316 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30421316 Leverandør Toronto Research Chemicals Leverandørnr. TRCP2305001MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Penicillin G-D7 N-Ethylpiperidinium Salt
CAS 1217445-37-8
Molekylær formel C16 2H7 H11 N2 O4 S . C7 H15 N
Mængde 1 mg
Synonym (2S,5R,6R)-6-[[2,2-Dideuterio-2-(2,3,4,5,6-pentadeuteriophenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid 1-Ethylpiperidine, 1-Ethylpiperidine (2S,5R,6R)-3,3-Dimethyl-7-oxo-6-(2-(phenyl-d5)acetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
InChI formel InChI=1S/C16H18N2O4S.C7H15N/c1-16(2)12(15(21)22)18-13(20)11(14(18)23-16)17-10(19)8-9-6-4-3-5-7-9;1-2-8-6-4-3-5-7-8/h3-7,11-12,14H,8H2,1-2H3,(H,17,19)(H,21,22);2-7H2,1H3/t11-,12+,14-;/m1./s1/i3D,4D,5D,6D,7D,8D2;
SMIL [2H]c1c([2H])c([2H])c(c([2H])c1[2H])C([2H])([2H])C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C(=O)O.CCN4CCCCC4
IUPAC navn (2S,5R,6R)-6-[[2,2-dideuterio-2-(2,3,4,5,6-pentadeuteriophenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;1-ethylpiperidine
Molekylvægt (g/mol) 454.634
Formel vægt 454.263