missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Penicillin Antibody (Pen-9), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14083AF488
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Penicillin Monoclonal antibody specifically detects Penicillin in All species samples. It is validated for ELISA, Immunofluorescence
Tekniske data
| Penicillin | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Ampicillin, Benzylpenicillin, dicloxacillin, flucloxacillin, methicillin, PCN, Penicilin, Penicillin G, Phenoxymethylpenicillin | |
| Penicilloyl-transferrin conjugate | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA, Immunofluorescence | |
| Pen-9 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| All species | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion