Få mere at vide
Penconazole-d7, TRC
Artikelnummer. 30416228
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30416228 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30416228 Leverandør Toronto Research Chemicals Leverandørnr. TRCP2218035MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Penconazole-d7
CAS 1628110-84-8
Molekylær formel C13 D7 H8 Cl2 N3
Mængde 5 mg
Synonym (±)-Penconazole-d7, CGA 71818-d7, Ofir-d7, Omnex-d7, Topas-d7
InChI formel InChI=1S/C13H15Cl2N3/c1-2-3-10(7-18-9-16-8-17-18)12-5-4-11(14)6-13(12)15/h4-6,8-10H,2-3,7H2,1H3/i1D3,2D2,3D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C(Cn1cncn1)c2ccc(Cl)cc2Cl
IUPAC navn 1-[3,3,4,4,5,5,5-heptadeuterio-2-(2,4-dichlorophenyl)pentyl]-1,2,4-triazole
Molekylvægt (g/mol) 291.23
Formel vægt 290.1082