Få mere at vide
PEN (mouse), Tocris Bioscience™
GPR83 agonist
Brand: Tocris Bioscience 6308/1
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
PEN (mouse) is a endogenous peptide GPR83 agonist. ProSAAS-derived neuropeptide. Activates phospholipase C (PLC)-mediated signaling cascade in mouse hypothalamus.Tekniske data
| 1236955-25-1 | |
| Store at -20°C | |
| Observed MW: 2317.62 | |
| 1 mg |