Få mere at vide
Pempidine Tartrate, LoGiCal
Artikelnummer. 30459918
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30459918 5 mg
1 options
Artikelnummer. 30459918 Leverandør LoGiCal Leverandørnr. LGCFOR1604.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pempidine Tartrate
CAS 546-48-5
Analyt- eller komponentnavne Pempidine Tartrate
Forsendelsestilstand Room Temperature
Molekylær formel C10 H21 N . C4 H6 O6
Certificeringer/compliance ISO 9001
Synonym Piperidine, 1,2,2,6,6-pentamethyl-, (2R,3R)-2,3-dihydroxybutanedioate (1:1)Piperidine, 1,2,2,6,6-pentamethyl-, [R-(R*,R*)]-2,3-dihydroxybutanedioate (1:1), Piperidine, 1,2,2,6,6-pentamethyl-, tartrate (1:1) (7CI,8CI), Piperidine, 1,2,2,6,6-pentamethyl-, tartrate (6CI), 1,2,2,6,6-Pentamethylpiperidine monotartrate, M and B 4486, Pempidil, Pempidine bitartrate, Pempidine tartrate, Pempiten, Perolysen, Sinapleg, Synapleg, Tenormal, Tensinol, Tensoral
InChI formel InChI=1S/C10H21N.C4H6O6/c1-9(2)7-6-8-10(3,4)11(9)5;5-1(3(7)8)2(6)4(9)10/h6-8H2,1-5H3;1-2,5-6H,(H,7,8)(H,9,10)/t;1-,2-/m.1/s1
Løsningstype Neat
SMIL CN1C(C)(C)CCCC1(C)C.O[C@H]([C@@H](O)C(=O)O)C(=O)O
IUPAC navn (2R,3R)-2,3-dihydroxybutanedioic acid;1,2,2,6,6-pentamethylpiperidine
Industry Type Clinical Testing
Molekylvægt (g/mol) 305.37
Mængde 5 mg
Formel vægt 305.1838 g/mol