Learn More
3M™ PELTOR™ WS™ LiteCom Headset
3M™ PELTOR™ WS™ LiteCom Headset takes wireless communication one step further by adding a Bluetooth™ link to the communication radio.
Brand: 3M™ 7100010979
This item is not returnable.
View return policy
Description
- Stay protected while taking calls wirelessly or listening to the radio in noisy environments
- Level dependent function lets the user hear surrounding noise and converse with colleagues
- Wireless Bluetooth™ connection to your mobile phone allows you to manage phone calls directly in the headset
- Wireless Bluetooth™ range up to 10 m (Class 2)
- Telephone calls can be managed simultaneously with a call on the communication radio
- Communication on 8 channels in the PMR 446 MHz band
- 38 sub channels allow users to communicate individually using the same channel
- Integrated noise cancelling microphone
- Automatically switches off after two hours of non-use to save batteries
Specifications
Headset | |
Neckband | |
MT53H7B4410WS5 | |
31 dB |
Product Content Correction
Product Title
3M™ PELTOR™ WS™ LiteCom Headset > Model: MT53H7B4410WS5
Spot an opportunity for improvement?Share a Content Correction