missing translation for 'onlineSavingsMsg'
Learn More

3M™ PELTOR™ FLX2 Cable for CH-3 Headset With External PTT Button

PELTOR™ FLX2 platform allows workers to pair protective communication CH-3 two-way headsets with an external push-to-talk button.

720.00 DKK

Specifications

Color Black
For Use With (Equipment) For use with CH-3 Headsets with external PTT Button
For Use With (Application) Enables connection to two-way communication for CH-3 Headsets
Pairs per Package 1 Ea.
View More Specs

Products 1
Product Code Brand Model Product Type Price Quantity & Availability  
Product Code Brand Model Product Type Price Quantity & Availability  
17861041
View Documents
3M™
7100185799
FLX2-200 FLX2 Cable J11
720.00 DKK
Each
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 30-09-2025
Add to basket
 
Description

Description

  • For use with CH-3 Headset with external PTT Button
  • J11 Plug Type
  • Coiled polyurethane cable
  • Aramid fiber reinforced
  • Robust and highly flexible

Compliance

3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

Specifications

Specifications

Black
For use with CH-3 Headsets with external PTT Button
Enables connection to two-way communication for CH-3 Headsets
1 Ea.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3M™ PELTOR™ FLX2 Cable for CH-3 Headset With External PTT Button

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.