Få mere at vide
Pelitinib-d6, TRC
Artikelnummer. 30421085
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30421085 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30421085 Leverandør Toronto Research Chemicals Leverandørnr. TRCP2187021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pelitinib-d6
CAS 1325223-34-4
Molekylær formel C24H17D6ClFN5O2
Mængde 1 mg
Synonym (2E)-N-[4-[(3-Chloro-4-fluorophenyl)amino]-3-cyano-7-ethoxy-6-quinolinyl]-4-(dimethylamino-d6)-2-butenamide, EKB 569-d6, WAY-EKB 569-d6
InChI formel InChI=1S/C24H23ClFN5O2/c1-4-33-22-12-20-17(11-21(22)30-23(32)6-5-9-31(2)3)24(15(13-27)14-28-20)29-16-7-8-19(26)18(25)10-16/h5-8,10-12,14H,4,9H2,1-3H3,(H,28,29)(H,30,32)/b6-5+/i2D3,3D3
SMIL FC(C=C1)=C(Cl)C=C1NC2=C(C#N)C=NC3=CC(OCC)=C(NC(/C=C/CN(C([2H])([2H])[2H])C([2H])([2H])[2H])=O)C=C32
Molekylvægt (g/mol) 473.96
Formel vægt 473.1901