Learn More
Thermo Scientific™ PEEK Fingertight Fittings
Product Code. 15461073 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Fingertight Fitting
One-Piece PEEK Fingertight Fitting
One-Piece PEEK Long Fingertight Fitting
PEEK Wing Nut w/PEEK Ferrule
Replacement Ferrule
Short Head Port Plug, Black
Short Head Port Plug, Red
Description:
1/16 in.
1/16 in., 0.37 in. Head
1/16 in., 10-32 Coned, Delrin, Black
1/16 in., 10-32 Coned, Delrin, Red
PEEK
PEEK, 1/32 in., 0.25 in. Head
Unit Size:
Each
Pack of 10
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Description Unit Size
15461073 Short Head Port Plug, Black 1/16 in., 10-32 Coned, Delrin, Black Each
10706163 One-Piece PEEK Fingertight Fitting 1/16 in., 0.37 in. Head Pack of 10
10754994 One-Piece PEEK Long Fingertight Fitting 1/16 in., 0.37 in. Head Pack of 10
10618915 Replacement Ferrule PEEK Pack of 10
11503980 PEEK Wing Nut w/PEEK Ferrule 1/16 in. Each
10504465 Fingertight Fitting PEEK, 1/32 in., 0.25 in. Head Pack of 10
15319849 Short Head Port Plug, Red 1/16 in., 10-32 Coned, Delrin, Red Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 15461073 Supplier Thermo Scientific™ Supplier No. U467BLK
Forventet forsendelse: 10-12-2026
Ensure HPLC quality and robustness with Thermo Scientific™ PEEK Fingertight Fittings.
Ensure HPLC quality and robustness with Thermo Scientific™ PEEK Fingertight Fittings. Machined for reliability, these fittings resist cracking, breaking, thread stripping and leaking in both low- and high-pressure applications. Fingertight plumbing ensures ease of use. The fittings are biocompatible for use across a broad range of applications.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Type Short Head Port Plug, Black
Description 1/16 in., 10-32 Coned, Delrin, Black
For Use With (Equipment) 10-32 Coned Port
Material Delrin Acetal
Quantity 1 Each