Få mere at vide
Pecazine Hydrochloride, TRC
Artikelnummer. 30421502
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30421502 100 mg
1 options
Artikelnummer. 30421502 Leverandør Toronto Research Chemicals Leverandørnr. TRCP217890100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pecazine Hydrochloride
CAS 738596-90-2
Molekylær formel C19H22N2S . xHCl
Mængde 100 mg
Synonym 10-[(1-Methyl-3-piperidyl)methyl]phenothiazine, III 2318, Lacumin, MPMP, Mepazin, Mepazine, Mepazine Base, Meprazine, Mesapin, Nothiazine, P 391, Pacatal, Pacatal base, Pacatol, Pakatal, Paxital, Pecatal, Pecazine
InChI formel InChI=1S/C19H22N2S.ClH/c1-20-12-6-7-15(13-20)14-21-16-8-2-4-10-18(16)22-19-11-5-3-9-17(19)21;/h2-5,8-11,15H,6-7,12-14H2,1H3;1H
SMIL CN1CC(CCC1)CN2C3=C(SC4=CC=CC=C42)C=CC=C3.Cl
IUPAC navn 10-((1-methylpiperidin-3-yl)methyl)-10H-phenothiazine hydrochloride
Molekylvægt (g/mol) 346.9