Learn More
BRAND™ LDPE Conical Ground Joint Stopper
LDPE construction with sealing rings. Octagonal grip provides easier grasping ability.
Brand: BRAND™ 144440
Additional Details : Weight : 0.01000kg
Description
- Low Density Polyethylene (LDPE) construction
- Hollow
- Octagonal grip
- With sealing rings
- Autoclavable to 121°C
Specifications
Conical bottom | |
1/Ea. | |
NS 34/35 | |
Octagonal |
Conical Ground Joint Stopper | |
LDPE | |
Yes |