missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ PDK4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA513776
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
PDK4 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
PDK4 (PDHK4), along with the other PDHKs, regulates glucose metabolism by phosphorylating the E1 alpha subunit of the mitochondrial pyruvate dehydrogenase complex. Inhibition of PDHKs is viewed as a potential therapeutic treatment for type II diabetes. This protein is located in the matrix of the mitrochondria and it's expression is regulated by glucocorticoids, retinoic acid and insulin. PDK4 inhibits the mitochondrial pyruvate dehydrogenase complex by phosphorylation of the E1 alpha subunit, thus contributing to the regulation of glucose metabolism.
Tekniske data
| PDK4 | |
| Polyclonal | |
| Unconjugated | |
| Pdk4 | |
| [Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 4, mitochondrial; [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 4, mitochondrial; AV005916; kinase isozyme 4; lipoamide; PDHK4; Pdk4; pyruvate dehydrogenase; pyruvate dehydrogenase kinase 4; Pyruvate dehydrogenase kinase isoform 4; pyruvate dehydrogenase kinase, isoenzyme 4; pyruvate dehydrogenase kinase, isozyme 4; pyruvate dehydrogenase, lipoamide, kinase isozyme 4, mitochondrial; pyruvate dehydrogenate kinase 4 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 27273, 5166 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O70571, Q16654 | |
| Pdk4 | |
| KLH conjugated synthetic peptide between 382-410 amino acids from the C-terminal region of human PDK4. | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion