missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PDK4 (PDKH4), GST Tag Recombinant Protein
Human PDK4 (PDKH4), GST Tag Recombinant Protein
Brand: Invitrogen PV5710
Additional Details : Weight : 0.00010kg
Description
PDK4 (PDHK4), along with the other PDHKs, regulates glucose metabolism by phosphorylating the E1 alpha subunit of the mitochondrial pyruvate dehydrogenase complex. Inhhibition of PDHKs is viewed as a potential therapeutic treament for type II diabetes.Specifications
NP_002603.1 | |
PDK4 | |
10 μg | |
PDK4 | |
Full-length | |
Recombinant | |
50 mM Tris pH 7.5, 150 mM NaCl, 0.02% Trition X-100, 0.5 mM EDTA, 2 mM DTT, 50% Glycerol. Kinase proteins are shipped on dry ice. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: Atypical | |
Dry Ice | |
N-Terminal | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |
Kinase Assay | |
73.9 kDa | |
PDK-4 (PDKH4) | |
PDHK Kinase Family | |
Human | |
GST Tag | |
10 μg | |
Human | |
Adapta™ Universal Kinase Assay | |
PDK4 | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |