Få mere at vide
PDE4A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA131151
Beskrivelse
PA1-31151 detects PDE4 from mouse, rat, human samples. PA1-31151 has been successfully used in Western blot applications. The PA1-31151 immunogen is: Synthetic peptide (unfortunately, the amino acid sequence is considered to be commercially sensitive) C terminal. Store at 4°C short term. For extended storage aliquot and store at -20°C or below. Avoid freeze-thaw cycles.
Cyclic nucleotides are important second messengers that regulate and mediate a number of cellular responses to extracellular signals, such as hormones, light, and neurotransmitters. Cyclic nucleotide phosphodiesterases (PDEs) regulate the cellular concentrations of cyclic nucleotides and thereby play a role in signal transduction. PDE4A is a class IV cAMP-specific PDE (summary by Milatovich et al., 1994 [PubMed 8009369]).
Tekniske data
| PDE4A | |
| Polyclonal | |
| Unconjugated | |
| P27815, O89084, P54748 | |
| PDE4A | |
| Synthetic peptide corresponding to the C terminal region. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| 0.68 mg/mL | |
| PDE4A | |
| DPDE2, PDE4, PDE46 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18577, 25638, 5141 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.