Få mere at vide
Tocris Bioscience™ PDD 00017272
GREENER_CHOICE
Artikelnummer. 18719886
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18719886 10 mg
18718156 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18719886 Leverandør Tocris Bioscience™ Leverandørnr. 7006/10
Potent PARG inhibitor
PDD 00017272 is a potent poly (ADP ribose) glycohydrolase (PARG) inhibitor (IC50 values are 4.8 nM in biochemical assay and 9.2 nM in cell based assay).
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-[(2,4-Dimethyl-5-thiazolyl)methyl]-1,2,3,4-tetrahydro-N-(1-methylcyclopropyl)-3-[(2-methyl-5-thiazolyl)methyl]-2,4-dioxo-6-quinazolinesulfonamide
CAS 1945950-20-8
Mængde 10 mg
Mål PARG Inhibitors
Molekylær formel C23H25N5O4S3
Renhed 98%