missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
STEMCELL Technologies PD0325901
Description
- PD0325901 is a selective, cell permeable inhibitor of the MEK/ERK pathway that inhibits the activation and downstream signaling of MEK
- It is an extremely potent inhibitor, suppressing the phosphorylation of ERK in C26 cells at very low concentrations (IC50 = 0.33 nM) (Bain et al.), Barrett et al.)
Specifications
Specifications
| CAS | 391210-10-9 |
| Mængde | 1 mg |
| Molekylær formel | C16H14F3IN2O4 |
| Renhed | ≥98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?