Få mere at vide
Tocris Bioscience™ PD 81723
GREENER_CHOICE
Artikelnummer. 18005192
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18005192 10 mg
18035182 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18005192 Leverandør Tocris Bioscience™ Leverandørnr. 1363/10
Allosteric potentiator of A1 receptors
Allosteric potentiator at the adenosine A1 receptor; acts via agonist-dependent and -independent mechanisms. Enhances agonist affinity for, and increased t½ of dissociation from, the receptor. Also inhibits basal and forskolin-stimulated adenylyl cyclase (AC) activity in A1 receptors expressed in CHO cells, possibly via direct potentiation of constitutive receptor activity or by direct inhibition of AC. Active in vivo.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 132861-87-1
Molekylær formel C14H12F3NOS
Tekniske data
Kemisk navn eller materiale PD 81723
Mængde 10 mg
Formel vægt 299.31
Renhed >99%
Molekylvægt (g/mol) 299.311