Få mere at vide
Tocris Bioscience™ PD 407824
GREENER_CHOICE
Artikelnummer. 18074092
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18074092 1 mg
18055262 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18074092 Leverandør Tocris Bioscience™ Leverandørnr. 2694/1
Selective inhibitor of Chk1 and Wee1
Selective inhibitor of checkpoint kinases Chk1 and Wee1 (IC50 values are 47 and 97 nM respectively). Displays selectivity over a range of other protein kinases; IC50 values are 3.4, 3.75, > 5, > 50, > 50 and > 50 μM for PKC, CDK4, other CDKs, c-Src, PDGFR and FGFR respectively.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 622864-54-4
Molekylær formel C20H12N2O3
Tekniske data
Kemisk navn eller materiale PD 407824
Mængde 1 mg
Formel vægt 328.32
Renhed >98%
Molekylvægt (g/mol) 328.33