Få mere at vide
R&D Systems™ PD 198306
Beskrivelse
Potent inhibitor of MEK1/2. Inhibits isolated enzyme at a concentration of 8 nM and inhibits MEK activity in synovial fibroblasts at concentrations of 30 - 100 nM. Highly selective for MEK; IC50 values are > 1, > 4, > 4 and > 10 μM for ERK, c-Src, cdks and PI 3-kinase γ respectively. Antihyperalgesic; blocks static allodynia in the streptozocin model of neuropathic pain following i.t. administration.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | PD 198306 |
| CAS | 212631-61-3 |
| Mængde | 10 mg |
| Produktlinje | Tocris |
| Molekylær formel | C18H16F3IN2O2 |
| Formel vægt | 476.23 |
| Renhed | >98% |
| Molekylvægt (g/mol) | 476.238 |