Få mere at vide
PD-166285-d4, TRC
Artikelnummer. 30427476
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30427476 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30427476 Leverandør Toronto Research Chemicals Leverandørnr. TRCP21750210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-(2,6-Dichlorophenyl)-2-((4-(2-(diethylamino)ethoxy-d4)phenyl)amino)-8-methylpyrido(2,3-d)pyrimidin-7(8H)-one Hydrochloride
CAS 1246814-59-4
Molekylær formel C26 D4 H23 Cl2 N5 O2
Mængde 10 mg
Synonym PD-166285-d4
InChI formel InChI=1S/C26H27Cl2N5O2/c1-4-33(5-2)13-14-35-19-11-9-18(10-12-19)30-26-29-16-17-15-20(25(34)32(3)24(17)31-26)23-21(27)7-6-8-22(23)28/h6-12,15-16H,4-5,13-14H2,1-3H3,(H,29,30,31)/i13D2,14D2
SMIL [2H]C([2H])(Oc1ccc(Nc2ncc3C=C(C(=O)N(C)c3n2)c4c(Cl)cccc4Cl)cc1)C([2H])([2H])N(CC)CC
IUPAC navn 6-(2,6-dichlorophenyl)-8-methyl-2-[4-[1,1,2,2-tetradeuterio-2-(diethylamino)ethoxy]anilino]pyrido[2,3-d]pyrimidin-7-one;dihydrochloride
Molekylvægt (g/mol) 516.456
Formel vægt 515.179