missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
PD-1-IN-17, MedChemExpress
Beskrivelse
PD-1-IN-17 is a programmed cell death-1 (PD-1) inhibitor extracted from patent WO2015033301A1, Compound 12, inhibits 92% splenocyte proliferation at 100 nM.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | PD-1-IN-17 |
| Fysisk form | Solid |
| CAS | 1673560-66-1 |
| Farve | White |
| Mængde | 50 mg |
| Renhedsgrad noter | Research |
| Molekylær formel | C13H22N6O7 |
| Til brug med (applikation) | Cancer-programmed cell death |
| Opløselighedsinformation | DMSO : 125 mg/mL (333.91 mM; Need ultrasonic) |
| SMIL | OC[C@@H](C(O)=O)NC(N[C@H](C1=NN=C([C@@H](N)[C@H](O)C)O1)CCC(N)=O)=O |
| Vis mere |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?