Få mere at vide
Tocris Bioscience™ PD 0332991 isethionate
GREENER_CHOICE
Artikelnummer. 30001693
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30001693 10 mg
30001694 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30001693 Leverandør Tocris Bioscience™ Leverandørnr. 4786/10
Potent, selective Cdk4/6 inhibitor; brain penetrant
Potent cyclin-dependent kinase 4 (cdk4) and cdk6 inhibitor (IC50 values are 11 nM and 15 nM respectively). Also displays activity at cdk2, cdk5 and cdk9 (IC50 values reported to be 1.6, 1.8 and 0.42-1.1 μM, respectively). Induces G1 cell cycle arrest and senescence in retinoblastoma protein (Rb)-proficient cell lines; blocks growth of intracranial glioblastoma multiforme xenografts in mice. Brain penetrant.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 827022-33-3
Molekylær formel C26H35N7O6S
Tekniske data
Kemisk navn eller materiale PD 0332991 isethionate
Analyseprocentområde >98%
Mængde 10 mg
Formel vægt 573.66
Molekylvægt (g/mol) 573.669