missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PBX2 Antibody (PCRP-PBX2-1C4), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14127AF700
Additional Details : Weight : 0.00970kg
Description
PBX2 Monoclonal antibody specifically detects PBX2 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
PBX2 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
5089 | |
Store at 4°C in the dark. | |
Purified |
Flow Cytometry, ELISA, Immunofluorescence | |
PCRP-PBX2-1C4 | |
Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
G17Homeobox protein PBX2, HOX12homeobox 12, PBX2MHC, pre-B-cell leukemia homeobox 2, pre-B-cell leukemia transcription factor 2, Protein G17 | |
Recombinant full-length human PBX2 protein (Uniprot: P40425) | |
0.1 mL | |
Primary | |
Human | |
Antibody | |
IgG2b |