Learn More
R&D Systems™ PBS, 100 Tablets
Product Code. 18769218
Change view
Click to view available options
Quantity:
1 Pk.
Unit Size:
1 set
Product Code. Quantity Unit Size
18769218 1 Pk. 1 set
1 options
Product Code. 18769218 Supplier R&D Systems™ Supplier No. 5564/1 Pack
  • ONLINE EXCLUSIVE PRICE
    1201.26 DKK
    1084.65 DKK / 1 sæt
    Spare 116.61 DKK
    9% Slukket
 Restricted Item
Forventet forsendelse: 25-11-2026
Add to basket
Edge
Add to basket
A Tocris product, part of R&D Systems. PBS Tablets
PBS, 100 Tablets is a phosphate buffered saline (PBS) tablets. Dissolve 1 tablet into 100ml of water. This will produce 100ml of PBS solution containing: Sodium chloride (137mM), Potassium chloride (2.7mM), Phosphate buffer (11.9mM).
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Pk.
Product Line Tocris
Target Buffers, Chelators and Reagents