Learn More
Alfa Aesar™ Paxilline, 97+%
Potent blocker of high-conductance calcium-activated potassium (BKCa) channels
Brand: Alfa Aesar™ J61341.MB
Additional Details : CAS Number : 57186-25-1 Weight : 0.00025kg Transport : UN number : 2811 Chem class : 6.1 Pack group : II
Quantity | 25mg |
---|
Chemical Identifiers
57186-25-1 | |
435.564 | |
ACNHBCIZLNNLRS-UBGQALKQSA-N | |
105008 | |
CC12CCC3C(=CC(=O)C(O3)C(C)(C)O)C1(CCC4C2(C5=C(C4)C6=CC=CC=C6N5)C)O |
C27H33NO4 | |
MFCD00083464 | |
paxilline, unii-3t9u9z96l7, 2h-1-benzopyrano 5',6':6,7 indeno 1,2-b indol-3 4bh-one, 5,6,6a,7,12,12b,12c,13,14,14a-decahydro-4b-hydroxy-2-1-hydroxy-1-methylethyl-12b,12c-dimethyl-, 2-alpha,4b-beta,6a-alpha,12b-beta,12c-alpha,14a-beta, 2h-pyrano 2,3:5',6' benz 1',2':6,7 indeno 1,2-b indol-3 4bh-one, 5,6,6a,7,12,12b,12c,13,14,14a-decahydro-4b-hydroxy-2-1-hydroxy-1-methylethyl-12b,12c-dimethyl-, 2r,4bs,6as,12bs,12cr,14as, spectrum5_001975, paxilline, powder hplc, paxilline solution, 100 mug/ml in acetonitrile, analytical standard, 2r,4bs,6as,12bs,12cr,14as-5,6,6a,7,12,12b,12c,13,14,14a-decahydro-4b-hydroxy-2-1-hydroxy-1-methylethyl-12b,12c-dimethyl-2h-pyrano 2,3:5',6' benz 1',2':6,7 indeno 1,2-b indol-3 4bh-one, ncgc00025342-07_c27h33no4_ 2r,4bs,6as,12bs,12cr,14as-4b-hydroxy-2-2-hydroxy-2-propanyl-12b,12c-dimethyl-5,6,6a,7,12,12b,12c,13,14,14a-decahydro-2h-chromeno 5',6':6,7 indeno 1,2-b indol-3 4bh-one | |
CHEBI:34907 |
Specifications
Paxilline | |
25mg | |
C27H33NO4 | |
UN2811 | |
Soluble in DMSO,acetone or chloroform. | |
CC12CCC3C(=CC(=O)C(O3)C(C)(C)O)C1(CCC4C2(C5=C(C4)C6=CC=CC=C6N5)C)O | |
105008 | |
435.56 | |
≥97% |
White to Yellow | |
57186-25-1 | |
MFCD00083464 | |
paxilline, unii-3t9u9z96l7, 2h-1-benzopyrano 5',6':6,7 indeno 1,2-b indol-3 4bh-one, 5,6,6a,7,12,12b,12c,13,14,14a-decahydro-4b-hydroxy-2-1-hydroxy-1-methylethyl-12b,12c-dimethyl-, 2-alpha,4b-beta,6a-alpha,12b-beta,12c-alpha,14a-beta, 2h-pyrano 2,3:5',6' benz 1',2':6,7 indeno 1,2-b indol-3 4bh-one, 5,6,6a,7,12,12b,12c,13,14,14a-decahydro-4b-hydroxy-2-1-hydroxy-1-methylethyl-12b,12c-dimethyl-, 2r,4bs,6as,12bs,12cr,14as, spectrum5_001975, paxilline, powder hplc, paxilline solution, 100 mug/ml in acetonitrile, analytical standard, 2r,4bs,6as,12bs,12cr,14as-5,6,6a,7,12,12b,12c,13,14,14a-decahydro-4b-hydroxy-2-1-hydroxy-1-methylethyl-12b,12c-dimethyl-2h-pyrano 2,3:5',6' benz 1',2':6,7 indeno 1,2-b indol-3 4bh-one, ncgc00025342-07_c27h33no4_ 2r,4bs,6as,12bs,12cr,14as-4b-hydroxy-2-2-hydroxy-2-propanyl-12b,12c-dimethyl-5,6,6a,7,12,12b,12c,13,14,14a-decahydro-2h-chromeno 5',6':6,7 indeno 1,2-b indol-3 4bh-one | |
ACNHBCIZLNNLRS-UBGQALKQSA-N | |
435.564 | |
CHEBI:34907 | |
Powder |