missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
PAX2 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20976-1-AP-20UL
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
The PAX2 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human PAX2. This antibody recognizes human, mouse, rat antigen. The PAX2 antibody has been validated for the following applications: WB, ELISA analysis.Tekniske data
| PAX2 | |
| Polyclonal | |
| Unconjugated | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 45 kDa | |
| Liquid |
| Western Blot, ELISA | |
| 450 μg/mL | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 5076 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion