Learn More
Emma Safety Footwear Patrick Safety Shoes
Product Code. 15998941
Change view
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15998941 47 1 pair
15998931 37 1 pair
15908941 38 1 pair
15918941 39 1 pair
15928941 40 1 pair
15938941 41 1 pair
15948941 42 1 pair
15958941 43 1 pair
15968941 44 1 pair
15978941 45 1 pair
15988941 46 1 pair
15908951 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 15998941 Supplier Emma Safety Footwear Supplier No. MM73084647
  • ONLINE EXCLUSIVE PRICE
    1140.62 DKK
    677.25 DKK / 1 par
    Spare 463.37 DKK
    40% Slukket
Forventet forsendelse: 16-03-2027
An ideal choice when optimal foot protection is required. A strong, solid steel plate in the sole and resilient leather uppers defend the feet from unexpected hazards. Perfectly balanced, anatomical arch support will keep feet feeling fit and fresh.
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • GripForce™ Duo Heat Resistant Outsole
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20345:2011
Closure Type Laces
Color Black
Design Patrick
Liner Material Hydro-Tec™ Sanitized Silver
Material Leather
Size 47
Toe Material Steel