missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ PATJ Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585204
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Keep as concentrated solution. Predicted reactivity: Bovine (100%). Positive Control: Mouse bladder. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein with multiple PDZ domains. PDZ domains mediate protein-protein interactions, and proteins with multiple PDZ domains often organize multimeric complexes at the plasma membrane. This protein localizes to tight junctions and to the apical membrane of epithelial cells. A similar protein in Drosophila is a scaffolding protein which tethers several members of a multimeric signaling complex in photoreceptors.
Tekniske data
| PATJ | |
| Polyclonal | |
| Unconjugated | |
| PATJ | |
| channel-interacting PDZ domain protein; channel-interacting PDZ domain-containing protein; Cipp; hINADL; inactivation no after-potential D-like protein; inactivation-no-afterpotential D-like; Inadl; Inadl protein; Inadl2; InaD-like; InaD-like (Drosophila); InaD-like 2; inaD-like protein; novel ankyrin-repeat protein; PALS1-associated tight junction protein; Patj; PATJ, crumbs cell polarity complex component; PDZ domain protein; protein associated to tight junctions; RGD1565362 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10207, 12695 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q63ZW7, Q8NI35 | |
| PATJ | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human PATJ. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion