Få mere at vide
DiscoverX PathHunter™ eXpress c-KIT Functional Assay
Mængde:
1.000 dp (10 x 96-brønds)
Beskrivelse
The PathHunter™ eXpress c-KIT Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated c-KIT dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cells overexpress PK-tagged c-KIT and an EA-tagged SH2 domain protein. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Cyno version of the protein.
Tekniske data
Tekniske data
| Assay Entry | Receptor Phosphorylation |
| Til brug med (applikation) | Assay-Ready eXpress Kits |
| Primær agonist | Recombinant Human SCF |
| Mængde | 1.000 dp (10 x 96-brønds) |
| Målindgang | c-KIT |
| Cellelinje | U2OS |
| Mål | c-KIT |
Korrektion af produktindhold
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.