Få mere at vide
DiscoverX PathHunter™ β-Arrestin2 Retroparticles
Artikelnummer. 17334369
Encompass
Skift visning
Mængde:
4 hætteglas x 0,5 ml
Artikelnummer. Mængde
17334369 4 hætteglas x 0,5 ml
1 options
Artikelnummer. 17334369 Leverandør DiscoverX Leverandørnr. 931087
 Begrænset vare
Forventet forsendelse: 07-12-2026
The PathHunter™ β-Arrestin2 Retroparticles are retroparticles for transducing cells with β-Arrestin2-EA to allow for arrestin recruitment assays. Contains 4 vials of cryopreserved retroparticles.
The PathHunter™ β-Arrestin2 Retroparticles are MMLV particles designed to transduce cells of interest with β-Arrestin2 fused to EA. In conjunction with a plasmid expressing a PK-tagged GPCR of interest, this allows for these cells to be used for β-Arrestin2 recruitment assays. This product uses EFC technology. Ligand induced recruitment of β-Arrestin2 to the activated GPCR leads to complementation and produces EFC signal. These retroparticles ship as 4 vials of cryopreserved retroparticles.
Tekniske data
Til brug med (applikation) Retroparticles
Produkttype Retroparticles
Mængde 4 hætteglas x 0,5 ml