missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pasteurella multocida Toxin (PMT) Detection Kit

Rapidly detect Pasteurella multocida toxin, the causal factor of progressive atrophic rhinitis in pigs with this assay.

Brand:  Thermo Scientific™ K000921-9

Additional Details : Weight : 0.01000kg


 View more versions of this product

Product Code. 13246089

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Detect Pasteurella multocida toxin, the causal factor of progressive atrophic rhinitis in pigs1, with Thermo Scientific™ Oxoid™ Pasteurella multocida Toxin PMT Kit. The enzyme-linked immunosorbent assay is designed to deliver specific, sensitive, and fast detection of Pasteurella multocida toxin in porcine samples.

Progressive atrophic rhinitis is a disease of pigs, caused by infection with toxigenic P. multocida.1 The pathogenesis of the disease is linked to a single, well-characterized component: the Pasteurella multocida toxin (PMT). The clinical manifestations of the disease are shortening or distortion of the snout, sneezing, nasal discharge, and epitaxis. Reduced growth rate is seen in severe cases.

Use ELISA Systems Pasteurella multocida Toxin PMT Detection Kit for the detection of Pasteurella multocida toxin in porcine samples.


  • Simple: Ready-to-use liquid reagents and concentrates make the assay easy to implement and run
  • Sensitive: Detect proteins at low levels for improved assurance of safety
  • Specific: Detect PMT in any type of sample, including extracts of primary mixed cultures obtained from, e.g., nasal swabs or washings2


Other names for the PMT include dermonecrotic toxin (DNT), heat labile toxin (HLT), turbinate atrophy toxin (TAT), and osteolytic toxin.

For Veterinary Use Only.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
Specifications

Specifications

Pasteurella multocida Toxin (PMT) Detection Kit
ELISA (Standard)
960 Tests (10 Plates)
Pasteurella multocida toxin
Conjugate, Microtitration plate, Positive control antigen, Reconstitution buffer, Sealing tape, Stop solution, Substrate, Wash buffer concentrate, White strips
2°C to 8°C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H360D-May damage the unborn child.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers