Learn More
3M™ Particulate Filters
3M™ Particulate Filters can be used with 3M™ Gas and Vapour Filters 6000 Series to add particulate protection (in conjunction with a 501 filter retainer), and are designed to optimise your field of vision. Our filters are lightweight and feature a bayonet-style connection.
Brand: 3M™ 7000037673
247.00 DKK valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 0.12600kg
Description
- Combines with the 6000 Series Gas Vapor cartridges to provide particulate and gas/vapor protection
- Note: 3M 501 filter retainers needed to combine the 5000 and 6000 series
Specifications
EN143 : 2000, CE Approved | |
P1R | |
Particulate Filter | |
6000 and 7000 Series face masks |
Filter retainer | |
Electret | |
Solid and liquid particles | |
Filter |