Learn More
3M™ Particulate Filter, 2000 Series
Offers low breathing resistance. The 3M Particulate Filter uses bayonet-style cartridges for half- or full-facemasks.
Brand: 3M™ 7100015050
538.86 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Lightweight particulate filters with low breathing resistance
- Bayonet-style cartridge connection for fitting to facepiece
- Filters approved to EN143:2000
Specifications
EN143 : 2000, CE Approved | |
Solid and liquid particles | |
Particulate Filter |
Bayonet | |
P2R | |
Half- and Full-face masks |
Applications and solutions series: Safety edition
File size: 7.7 MB
Water analysis
File size: 7.3 MB
Product Content Correction
Product Title
3M™ Particulate Filter, 2000 Series > Filter type: P2R
Spot an opportunity for improvement?Share a Content Correction