Få mere at vide
Parkins’ Catalyst, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Parkins' Catalyst |
| CAS | 173416-05-2 |
| Molekylær formel | C6 H21 O3 P3 Pt |
| Mængde | 10 mg |
| Synonym | Ghaffar-Parkins Catalyst, Hydrido(dimethylphosphinous acid-kP)[hydrogen bis(dimethylphosphinito-kP)]platinum (II) Ghaffar-Parkins Catalyst |
| InChI formel | InChI=1S/3C2H7OP.Pt.H/c3*1-4(2)3;;/h3*3H,1-2H3;; |
| SMIL | C[P](C)(O)[PtH]([P](C)(C)O)[P](C)(C)O |
| Molekylvægt (g/mol) | 429.228 |
| Formel vægt | 429.035 |